Learn More
LIFE TECH (BID ECHO ONLY) E-Gel™ 48 Agarose Gels, 1%, 8 Gels
Supplier:  LIFE TECH (BID ECHO ONLY) G800801
Encompass
Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.
Catalog No. NC3859526