Learn More
Medchemexpress LLC (E)-JP-2-196 TFA | 3065021-33-9 | 97.9% | 388.34 | C17H19F3N2O5 | 50mg
SDP
Supplier:  Medchemexpress LLC HYW854844A50MG
Encompass_Preferred
JP-2-196 (TFA) is a molecular glue handle that can bind to the E3-ubiquitin ligases (RNF126 LRSAM1 RNF40 MID2 and RNF14) in vitro JP-2-196 (TFA) can be used to synthesize molecular glue degraders[1]
Catalog No. 50-002-70239