Learn More
Medchemexpress LLC HY-N2492 100mg Medchemexpress, (E)-Methyl 4-coumarate CAS:19367-38-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2492100MG
Encompass_Preferred
Medchemexpress, HY-N2492 100mg (E)-Methyl 4-coumarate CAS:19367-38-5 (E)-Methyl 4-coumarate (Methyl 4-hydroxycinnamate), found in several plants, such as green onion (Allium cepa) or noni (Morinda citrifolia L.) leaves. (E)-Methyl 4-coumarate cooperates with Carnosic Acid in inducing apoptosis and killing acute myeloid leukemia cells, but not normal peripheral blood mononuclear cells. Antioxidant and antimicrobial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2900