Learn More
Medchemexpress LLC (E)-N-(4-(1-(3-Aminobenzoyl)piperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide | 2222981-61-3 | 99.5% | 406.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY40223710MM/1ML
Encompass_Preferred
Nampt-IN-15 is an inhibitor of Nampt (nicotinamide phosphoribosyltransferase). It exhibits cytotoxicity against various cell lines, including BxPC-3, HepG2, L540cy, and MOLM-13, with IC50 values ranging from 7 nM to 38.5 nM. The product is intended for research use only and is not for sale to patients.
  • Functions as an inhibitor of Nampt.
  • Demonstrates cytotoxicity against BxPC-3, HepG2, L540cy, and MOLM-13 cell lines.
  • Available in solid form.
  • Provided with a data sheet, certificate of analysis (COA), and safety data sheet (SDS).
  • Shipping at room temperature in continental US; storage at 4°C, protected from light.
  • Stock solutions in solvent can be stored at -80°C for 6 months or -20°C for 1 month, protected from light.
Catalog No. 50-004-01598