Learn More
Medchemexpress LLC (E)-N-((Z)-4-benzyl-3-methyl-1,2,4-thiadiazol-5(4H)-ylidene)-N'-phenylbenzimidamide | 324579-65-9 | 99.9% | 384.50 g/mol | C23H20N4S | 10MG

Supplier: Medchemexpress LLC HY12396710MG
RNF5 inhibitor inh-02 is a small-molecule inhibitor of the E3 ubiquitin ligase RNF5 (RMA1) that reduces ATG4B ubiquitylation, increases LC3IIB expression and autophagic vacuole number, and rescues F508del-CFTR function in cellular models (reported EC50 ≈ 2.2-2.6 μM). It is supplied as a solid or DMSO solution for research use in cellular assays and mechanistic studies.
- High purity (99.91%).
- Molecular formula C23H20N4S and molecular weight 384.50 g/mol.
- Demonstrated cellular activity rescuing F508del-CFTR (EC50 ≈ 2.2-2.6 μM).
- Soluble in DMSO (~83.33 mg/mL); ultrasonic heating to 60°C recommended for dissolution.
- Available as solid and as prepared DMSO solution formats for convenience.
- Suitable for research into RNF5 biology and cystic fibrosis-related cellular models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.