Learn More
Medchemexpress LLC (E)-p-Coumaryl alcohol | 20649-40-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN3102A5MG
Encompass_Preferred
(E)-p-Coumaryl alcohol is a metabolite of coumarin with significant cytotoxicity. It can be isolated from Alpinia officinarum and Rhodiola rosea and is used for inflammation research.
  • Metabolite of coumarin
  • Exhibits significant cytotoxicity
  • Isolated from Alpinia officinarum and Rhodiola rosea
  • Suitable for inflammation research
  • Has a purity of 99.50%
  • Appears as a white to off-white solid
Catalog No. 50-004-21221