Learn More
Medchemexpress LLC (E)-p-Coumaryl alcohol | 20649-40-5 | 50 MG

Supplier: Medchemexpress LLC HYN3102A50MG
(E)-p-Coumaryl alcohol is a metabolite of coumarin with significant cytotoxicity. It can be isolated from Alpinia officinarum and Rhodiola rosea and is used for inflammation research.
- Significant cytotoxicity
- Isolated from Alpinia officinarum and Rhodiola rosea
- Used for inflammation research
- Purity of 99.50%
- Molecular weight of 150.17
- Molecular formula is C9H10O2
- Appearance is solid
- Color is white to off-white
- Shipping at room temperature (continental US)
- Storage at -20°C, protected from light
- Soluble in DMSO: 125 mg/mL (requires ultrasonication)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.