Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-p-coumaryl alcohol | 20649-40-5 | MFCD02169461 | 99.5% | 150.17 | C9H10O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN3102A10MG

Encompass_Preferred

(E)-p-Coumaryl alcohol is a naturally occurring coumarin metabolite used in research on inflammation and cytotoxicity. It is supplied as a white to off-white solid for laboratory use.

  • Purity 99.5%.
  • CAS number 20649-40-5.
  • Molecular formula C9H10O2.
  • Molecular weight 150.17 g/mol.
  • Appearance white to off-white solid.
  • Storage solid: keep at -20°C and protect from light; in solvent: store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-89202


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.