Learn More
Medchemexpress LLC E -Pent-2-enal 500g
SDP
Supplier:  Medchemexpress LLC HYW111375500G
Encompass_Preferred
(E)-Pent-2-enal has a pungent fruity odor This compound is commonly used in the flavor and fragrance industry because of its strong aroma often described as fresh and green Furthermore (E)-Pent-2-enal can be used as an intermediate in the synthesis of various organic compounds including pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in many commercial products including perfumes air fresheners and cleaners
Catalog No. 50-002-04987