Learn More
Medchemexpress LLC E-Selectin/CD62E Hu 100ug
SDP
Supplier:  Medchemexpress LLC HYP74161100UG
Encompass_Preferred
E-selectin/CD62E Protein a cell-surface glycoprotein crucially mediates immunoadhesion by interacting with SELPLG/PSGL1 for the adhesion of neutrophils to cytokine-activated endothelium It also potentially influences capillary morphogenesis and interacts with SELPLG/PSGL1 and PODXL2 through the sialyl Lewis X epitope independently of SELPLG sulfation E-Selectin/CD62E Protein Human (HEK293 His-Fc) is the recombinant human-derived E-Selectin/CD62E protein expressed by HEK293 with C-hFc C-His labeled tag
Catalog No. 50-001-79444