Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E-selectin (CD62E) protein, cynomolgus (HEK293, His) | 98.0% | 5 UG
SDP

Supplier:  Medchemexpress LLC HYP752205UG

Encompass_Preferred

Recombinant E-selectin (CD62E) protein from cynomolgus macaque expressed in HEK293 cells. The product includes a C-terminal His tag and is intended for in vitro adhesion and binding studies.

  • Recombinant cynomolgus E-selectin protein.
  • Expressed in HEK293 cells.
  • C-terminal His tag for detection or purification.
  • Purity 98.0%.
  • Available sizes include 5 UG, 10 UG, 50 UG.
  • Intended for cell adhesion and endothelial interaction assays.

Catalog No. 50-002-92905


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.