Learn More
Medchemexpress LLC E Z -4 4 -Dicyanost 100mg
SDP
Supplier:  Medchemexpress LLC HYW112166100MG
Encompass_Preferred
(E/Z)-4 4 -Dicyanostilbene is the isomer of 4 4 -Dicyanostilbene (HY-W112166A) and can be used as an experimental control 4 4 -Dicyanostilbene (compound 43) is a potent antimalarial agent against the Dd2 strain with an EC50 of 27 nM 4 4 -Dicyanostilbene exhibits in vivo efficacy against methicillin-resistant Staphylococcus aureus (MRSA)[1]
Catalog No. 50-002-06519