Learn More
Medchemexpress LLC (E/Z)-4-Hydroxytamoxifen (Afimoxifene) | 68392-35-8 | 99.62% | 387.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16950A100MG
Encompass_Preferred
(E/Z)-4-Hydroxytamoxifen (Afimoxifene) is a racemic compound comprised of (Z)-4-Hydroxytamoxifen and (E)-4-Hydroxytamoxifen isomers. It functions as a selective estrogen receptor modulator, exhibiting both estrogenic and antiestrogenic properties. It is also an active metabolite of Tamoxifen. The compound shows promise for research related to cyclical mastalgia, including breast pain, tenderness, and nodularity.
  • Functions as a selective estrogen receptor modulator
  • Exhibits both estrogenic and antiestrogenic properties
  • Active metabolite of tamoxifen
  • Acts as an agonist of the G protein-coupled estrogen receptor (GPER) with relatively low affinity (100-1000 nM)
  • Shows promise for research related to cyclical mastalgia, including breast pain, tenderness, and nodularity
Catalog No. 50-003-91118