Learn More
TARGETMOL CHEMICALS INC E/Z-BIX02188 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T63245MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIX02188 is a specific MEK5 inhibitor (IC50 4.3 nM) also inhibits ERK5 catalytic activity (IC50 810 nM) and does not inhibit closely related kinases MEK1/2 JNK2 and ERK2. purity: 99%
Catalog No. 50-257-6530