Learn More
Medchemexpress LLC (E/Z)-CCR-11 | 301687-87-6 | 98.0% | 5 MG

Supplier: Medchemexpress LLC HY119648A5MG
(E/Z)-CCR-11 is a small-molecule inhibitor of CD38 cyclase that increases cellular NAD+ levels and interferon γ. It has an IC50 of 20.8 μM against CD38 and is supplied as a high-purity research compound.
- Selective CD38 inhibitor (IC50 20.8 μM).
- Promotes increases in cellular NAD+ and interferon γ.
- Purity 98.0%.
- Molecular weight 355.35 g·mol-1; chemical formula C15H8F3NO2S2.
- Soluble in DMSO; 100 mg/mL with ultrasonic assistance.
- Recommended storage: powder -20 °C (3 years) or 4 °C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.