Learn More
Medchemexpress LLC (E/Z)-demethoxycurcumin (p-hydroxycinnamoyl-feruloylmethane) | 24939-17-1 | 98.0% | 338.35 g·mol⁻¹ | C20H18O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0006A10MG
Encompass_Preferred
(E/Z)-Demethoxycurcumin is a curcuminoid isolated from Curcuma species, described as having anticoagulative activity. Supplied as a high-purity solid for research use, it is suitable for biochemical, pharmacological, and antioxidant studies.
  • High purity suitable for research applications.
  • Curcuminoid with reported anticoagulative activity.
  • Applicable to anti-inflammatory and antioxidant studies.
  • Solid form; store at 4°C and protect from light.
  • Provided in small-scale quantities for laboratory experiments.
Catalog No. 50-002-97685