Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-E64FC26 | 2285446-67-3 | 99.8% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY122895A10MM 1ML

Encompass_Preferred

E/Z-E64FC26 is a mixture of E and Z stereoisomers used as a cysteine protease and apoptosis pathway inhibitor for research applications. It is supplied as a high-purity solid or as a 10 mM solution in DMSO for biochemical assays.

  • Mixture of E and Z stereoisomers.
  • Used as a cysteine protease and apoptosis inhibitor.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid quantities.
  • High purity: 99.8%.
  • Molecular weight 340.38 g/mol; formula C19H23F3O2.
  • Appearance: light yellow solid.

Catalog No. 50-002-31028


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.