Learn More
Medchemexpress LLC HY-122895A 5mg Medchemexpress, (E/Z)-E64FC26 CAS:2285446-67-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY122895A5MG
Encompass_Preferred
Medchemexpress, HY-122895A 5mg (E/Z)-E64FC26 CAS:2285446-67-3 (E/Z)-E64FC26 is a mixture complex of E-E64FC26 and Z-E64FC26. E64FC26 (E-E64FC26) is a potent pan-inhibitor of the protein disulfide isomerase (PDI) family, with IC50s of 1.9, 20.9, 25.9, 16.3, and 25.4 μM against PDIA1, PDIA3, PDIA4, TXNDC5, and PDIA6. E64FC26 shows anti-myeloma activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9938