Learn More
Medchemexpress LLC E/Z-GO289 10mM 1mL | 694522-87-7 | 419.30 g/mol | C17H15BrN4O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11551910MM 1ML
Encompass_Preferred
(E/Z)-GO289 is a potent, selective inhibitor of casein kinase 2 (CK2) used in biochemical and cell-based research. Supplied as a ready-to-use 10 mM solution in DMSO, it exhibits low-nanomolar activity and high stated purity appropriate for assay work.
  • Potent CK2 inhibition with IC50 ≈ 7 nM.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • High stated purity (~98.98%) suitable for biochemical assays.
  • Demonstrated to lengthen circadian period in cellular studies.
  • Appropriate for cell-based and biochemical research applications.
Catalog No. 50-002-29769