Learn More
Medchemexpress LLC (E/Z)-GSK-3β inhibitor 1 | 3367-88-2 | 99.1% | 222.24 | C14H10N2O | 50MG

Supplier: Medchemexpress LLC HY126144A50MG
(E/Z)-GSK-3β inhibitor 1 is a small-molecule research compound used to inhibit glycogen synthase kinase-3β (GSK-3β) in biochemical and cellular studies. The component compound shows potent activity and is provided as a high-purity solid with recommended storage and solvent stability information for maintaining sample integrity during research applications.
- Potent GSK-3β inhibition (IC50 = 4.9 nM).
- Reported purity 99.1%.
- Molecular formula C14H10N2O; molecular weight 222.24.
- Solid, orange to red appearance.
- Store at -20°C protected from light; in solvent: -80°C for up to 6 months, -20°C for 1 month.
- Supplied in multiple pack sizes for research use.
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.