Learn More
Medchemexpress LLC (E/Z)-GSK5182 | 2699724-40-6 | 98.9% | 417.54 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY111226A10MG
Encompass_Preferred
(E/Z)-GSK5182 is a racemic compound of (E)-GSK5182 and (Z)-GSK5182 isomers. GSK5182 is a highly selective and orally active inverse agonist of estrogen-related receptor γ (ERRγ) with an IC50 of 79 nM. It also induces reactive oxygen species (ROS) generation in hepatocellular carcinoma.
  • Racemic compound of (E)-GSK5182 and (Z)-GSK5182 isomers
  • Highly selective and orally active inverse agonist of estrogen-related receptor γ (ERRγ)
  • IC50 of 79 nM for ERRγ
  • Induces reactive oxygen species (ROS) generation in hepatocellular carcinoma
  • For research use only
Catalog No. 50-003-43102