Learn More
Medchemexpress LLC (E/Z)-Icerguastat | 13098-73-2 | 99.4% | 196.64 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1895610MM/1ML
Encompass_Preferred
(E/Z)-Icerguastat is a selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A). It is utilized for research into protein misfolding diseases.
  • Selective inhibitor of phosphatase regulatory subunit PPP1R15A
  • Used in protein misfolding diseases research
  • For research use only
Catalog No. 50-003-85109