Learn More
Medchemexpress LLC (E/Z)-Icerguastat ((E/Z)-Sephin1) | 13098-73-2 | 99.4% | 196.64 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18956
Encompass_Preferred
(E/Z)-Icerguastat ((E/Z)-Sephin1) is a selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A). It can be used for protein misfolding diseases research.
  • Selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A).
  • Applicable for research in protein misfolding diseases.
Catalog No. 50-003-85933