Learn More
Medchemexpress LLC (E/Z)-IT-603 | 292168-90-2 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HY12150810MM 1ML
(E/Z)-IT-603 is a mixture of E-IT-603 and Z-IT-603 that acts as an IκBα/c-Rel inhibitor and is supplied as a 10 mM solution in DMSO for research use.
- Provided as a 1 mL solution in DMSO.
- Concentration 10 mM in DMSO.
- Purity 99.9%.
- Molecular weight 329.17 g/mol; formula C11H9BrN2O3S.
- CAS number 292168-90-2.
- Reported c-Rel IC50 approximately 3 μM.
- Store in solution at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.