Learn More
Medchemexpress LLC HY-121508 5mg Medchemexpress, (E/Z)-IT-603 CAS:292168-90-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1215085MG
Encompass_Preferred
Medchemexpress, HY-121508 5mg (E/Z)-IT-603 CAS:292168-90-2 (E/Z)-IT-603 is a mixture of E-IT-603 and Z-IT-603 (IT-603). IT-603 is a c-Rel inhibitor with an IC50 of 3 μM. IT-603 has anti-tumor activity. (E/Z)-IT-603 is a promising modulator of T-cell responses in the context of graft-versus-host disease (GVHD) and malignant diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0969