Learn More
Medchemexpress LLC (E/Z)-J147 | 1146963-51-0 | 99.54% | 350.34 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1377950MG
Encompass_Preferred
(E/Z)-J147 is an exceptionally potent, orally active, neuroprotective agent designed for cognitive enhancement. It can readily cross the blood-brain barrier (BBB) and holds potential for research into Alzheimer's disease (AD). This product is intended for research use only.
  • Inhibits monoamine oxidase B (MAO B) and the dopamine transporter
  • Promotes survival of HT22 and primary cells
  • Enhances memory and dendritic spine number
  • Reverses reduction in recognition index
Catalog No. 50-003-49619