Learn More
Medchemexpress LLC E/Z-Locostatin | 133812-16-5 | 99.52% | 245.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01341150MG
Encompass_Preferred
E/Z-Locostatin is a racemic mixture of Locostatin. This potent RKIP inhibitor binds to Raf kinase inhibitor RKIP protein, disrupting its interaction with Raf-1 kinase and G protein-coupled receptor kinase 2. It inhibits cell proliferation and migration.
  • Potent RKIP inhibitor
  • Disrupts interaction of RKIP with Raf-1 kinase and G protein-coupled receptor kinase 2
  • Inhibits cell proliferation
  • Inhibits cell migration
  • Aggravates thioacetamide-induced acute liver failure in mice
Catalog No. 50-004-54570