Learn More
Medchemexpress LLC E/Z-mirin | 299953-00-7 | 99.2% | 220.25 g/mol | C10H8N2O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11769310MM 1ML
Encompass_Preferred
E/Z-Mirin is a stereoisomeric mixture of (E)- and (Z)-mirin that inhibits the MRN (Mre11-Rad50-Nbs1) complex and prevents MRN-dependent ATM activation, used in research on DNA damage response and cell cycle signaling.
  • Inhibits MRN complex and blocks MRN-dependent ATM activation
  • Supplied as a 10 mM solution in DMSO and as solid quantities for flexible dosing
  • High purity suitable for biochemical and cell-based assays
  • Low molecular weight (220.25 g/mol) facilitates cellular studies
  • SMILES provided for cheminformatics and compound registries
Catalog No. 50-002-26437