Learn More
Medchemexpress LLC (E/Z)-mirin | 299953-00-7 | MFCD05885480 | 99.2% | 220.25 | C10H8N2O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117693100MG
Encompass_Preferred
(E/Z)-Mirin is a research-grade small molecule inhibitor of the Mre11-Rad50-Nbs1 (MRN) complex that blocks MRN-dependent ATM activation and is used to study DNA damage response and ATM signaling.
  • Inhibits MRN-dependent ATM activation.
  • Tool compound for DNA damage response and cell cycle studies.
  • High purity (99.22%).
  • Molecular weight 220.25 and formula C10H8N2O2S.
  • Supplied as a powder and as ready-to-use DMSO solution formats.
  • Commonly supplied in a 100 MG pack size.
Catalog No. 50-002-27909