Learn More
Medchemexpress LLC (E/Z)-MitoTam iodide, hydriodide | 98.6% | 999.82 g·mol⁻¹ | C52H60I2NOP | 5 MG
SDP
Supplier:  Medchemexpress LLC HY126222B5MG
Encompass_Preferred
(E/Z)-MitoTam iodide, hydriodide is the E/Z isomeric mixture of a tamoxifen-derived mitochondrial electron transport chain inhibitor used in research to disrupt mitochondrial respiration, alter mitochondrial morphology, and induce apoptosis in cancer and senescent cells.
  • Inhibits mitochondrial electron transport chain and complex I respiration.
  • Reduces mitochondrial membrane potential in targeted cells.
  • Induces apoptosis in various breast cancer cell lines.
  • Eliminates senescent cells in vivo and reduces senescence markers.
  • Provided at high purity for research applications (≈98.6%).
Catalog No. 50-002-64847