Learn More
Medchemexpress LLC E/Z-necrosulfonamide | 432531-71-0 | MFCD02370191 | 98.8% | 461.47 g·mol⁻1 | C18H15N5O6S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100573A25MG
Encompass_Preferred
(E/Z)-Necrosulfonamide is the racemic form of necrosulfonamide, a small-molecule inhibitor that targets MLKL and Gasdermin D to block necroptosis and pyroptosis. It is supplied as a powder for research use in cell-death and inflammation studies.
  • Inhibits MLKL and Gasdermin D, blocking necroptosis and pyroptosis.
  • Useful for mechanistic studies of programmed cell death pathways.
  • Supplied as a stable powder suitable for long-term storage.
  • Reported high purity supports reproducible biochemical assays.
  • Available in small research-scale quantities for screening and validation.
Catalog No. 50-002-12680