Learn More
Medchemexpress LLC (E/Z)-Polydatin ((E/Z)-Piceid) | 65914-17-2 | 98.13% | 390.38 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN012010MM/1ML
Encompass_Preferred
(E/Z)-Polydatin ((E/Z)-Piceid) is a mixture of the E/Z configurations of Polydatin (HY-N0120A). Polydatin can be isolated from Polygonum cuspidatum, grapes, peanuts, red wine, hop pellets, cocoa-containing products, and chocolate products. It exhibits multiple biological properties and shows favorable cytotoxic effects against various tumor cell lines.
  • Exhibits anti-platelet aggregation
  • Anti-low-density lipoprotein oxidation properties
  • Cardioprotective activity
  • Anti-inflammatory and immunomodulatory functions
  • Shows cytotoxic effects against cervical cancer cells, liver cancer cells, and nasopharyngeal carcinoma cells
Catalog No. 50-004-30952