Learn More
Medchemexpress LLC (E/Z)-Sib-1893 | 6266-99-5 | 99.8% | 195.26 g·mol^-1 | C14H13N | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1020945MG
Encompass_Preferred
(E/Z)-SIB-1893 is the racemic mixture of the (E)- and (Z)-isomers of SIB-1893, a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5). Supplied as a solid research reagent, it is used in pharmacological and neurophysiological studies to probe mGluR5-mediated signaling. Molecular formula: C14H13N. Molecular weight: 195.26 g·mol^-1. Typical purity: 99.8%.
  • Racemic mixture of (E)- and (Z)-isomers for comparative pharmacology
  • Selective, non-competitive mGluR5 antagonist for receptor research
  • High stated purity (~99.8%) suitable for assay work
  • Supplied as a solid in small quantities for screening and method development
  • Also available in solution formats for cell-based and biochemical assays
Catalog No. 50-002-48513