Learn More
Medchemexpress LLC (E/Z)-Sib-1893 | 6266-99-5 | 99.8% | 195.26 g/mol | C14H13N | 100 MG

Supplier: Medchemexpress LLC HY102094100MG
(E/Z)-SIB-1893 is the racemic mixture of the (E)- and (Z)-isomers of SIB-1893, a selective non-competitive antagonist of metabotropic glutamate receptor subtype 5 (mGluR5) used as a research tool in neuroscience and pharmacology. Supplied as a solid powder and as a ready-to-use 10 mM solution in DMSO.
- Racemic mixture of (E)- and (Z)-isomers
- Selective non-competitive mGluR5 antagonist for research applications
- CAS number 6266-99-5
- Molecular formula C14H13N; molecular weight 195.26 g/mol
- High purity (99.8% assay)
- Available as solid powder and 10 mM solution in DMSO
- Common pack size: 100 mg; larger sizes available by request
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.