Learn More
Medchemexpress LLC (E/Z)-SIB-1893 | 6266-99-5 | MFCD02262120 | 99.8% | 195.26 | C14H13N | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10209450MG
Encompass_Preferred
(E/Z)-SIB-1893 is a racemic research compound consisting of the (E)- and (Z)-isomers of SIB-1893. The (E)-isomer is described as a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5). The product is supplied for laboratory research as a solid (50 mg) with listed purity 99.8% (CAS 6266-99-5), molecular formula C14H13N, and molecular weight 195.26.
  • Racemic mixture of (E)- and (Z)-isomers.
  • (E)-isomer acts as a selective non-competitive mGluR5 antagonist.
  • High listed purity suitable for research applications.
  • Available as a solid; solution formulations are offered separately.
Catalog No. 50-002-46192