Learn More
Medchemexpress LLC (E/Z)-Sivopixant | 1640808-39-4 | 98.6% | 507.93 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY137451A10MM/1ML
Encompass_Preferred
(E/Z)-Sivopixant is a potent P2X3 receptor antagonist with an IC50 of 4 nM. It is primarily used for respiratory diseases research. P2X3 receptors, expressed in neuroepithelial bodies of the lungs, are involved in information transmission in respiratory organs, including ATP-induced cough.
  • Potent P2X3 receptor antagonist
  • IC50 of 4 nM
  • Used for respiratory diseases research
  • Inhibits ATP-induced cough
Catalog No. 50-003-52265