Learn More
Medchemexpress LLC (E/Z)-Teriflunomide | 108605-62-5 | 99.87% | 270.21 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11015950MG
Encompass_Preferred
(E/Z)-Teriflunomide is the active metabolite of Leflunomide and functions as an immunomodulatory agent. It exerts its effects by inhibiting the mitochondrial enzyme dihydroorotate dehydrogenase (DHODH), which plays a crucial role in pyrimidine synthesis. This compound also acts as a non-steroidal anti-inflammatory drug and a tyrosine kinase inhibitor. It is utilized in the treatment of relapsing forms of multiple sclerosis and rheumatoid arthritis.
  • Active metabolite of Leflunomide
  • Immunomodulatory agent
  • Inhibits mitochondrial enzyme dihydroorotate dehydrogenase (DHODH)
  • Functions as a non-steroidal anti-inflammatory drug
  • Functions as a tyrosine kinase inhibitor
  • Inhibits pyrimidine synthesis
  • Used to treat relapsing forms of multiple sclerosis
  • Used to treat rheumatoid arthritis
Catalog No. 50-003-44244