Learn More
Medchemexpress LLC (E/Z)-tyrphostin AG490 | 134036-52-5 | 99.7% | 294.30 g·mol⁻¹ | C17H14N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10745925MG
Encompass_Preferred
(E/Z)-AG490 is the racemic mixture of the E and Z isomers of tyrphostin AG490, a small-molecule tyrosine kinase inhibitor commonly used in research to probe EGFR, JAK2/3, and STAT3 signaling. Provided as a high-purity solid, it is suitable for biochemical and cellular assays; powder storage at -20°C preserves stability, with specific solvent storage guidance for solutions.
Catalog No. 50-002-39753