Learn More
Medchemexpress LLC (E/Z)-ZINC09659342 | 591726-26-0 | 99.7% | C23H15F3N2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY145915A10MM/1ML
Encompass_Preferred
(E/Z)-ZINC09659342 is an inhibitor of Lbc-RhoA interaction. This compound appears as a solid with an orange to red color and has a purity of 99.71%. It is intended for research use only.
  • Inhibits Lbc-RhoA interaction
  • Solid form, orange to red color
  • Soluble in DMSO (≥ 50 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-54067