Learn More
Medchemexpress LLC (E/Z)-ZINC09659342 | 591726-26-0 | 99.7% | C23H15F3N2O4 | 25 MG

Supplier: Medchemexpress LLC 25MGHY145915A
(E/Z)-ZINC09659342 is an inhibitor of Lbc-RhoA interaction. It is provided as an orange to red solid with a purity of 99.7% and is intended for research use only. This compound targets Ras and is involved in the GPCR/G Protein and MAPK/ERK pathways.
- Inhibits Lbc-RhoA interaction
- Targets Ras and related pathways (GPCR/G Protein, MAPK/ERK)
- Molecular formula: C23H15F3N2O4
- Molecular weight: 440.37
- Purity: 99.7%
- Solubility in DMSO: ≥ 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.