Learn More
Medchemexpress LLC (E/Z)-Zotiraciclib ((E/Z)-TG02) | 937270-47-8 | C23H24N4O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1516650MG
Encompass_Preferred
(E/Z)-Zotiraciclib is a potent inhibitor of CDK2, JAK2, and FLT3, with IC50s of 13, 73, and 56 nM respectively. It effectively inhibits the proliferation of cancer cells and is suitable for cancer research.
  • Potent inhibitor of CDK2, JAK2, and FLT3.
  • Effectively inhibits proliferation of cancer cells.
  • Inhibits tumor growth in animal models.
  • High solubility in DMSO.
  • Suitable for cancer research.
Catalog No. 50-003-72950