Learn More
Medchemexpress LLC (E/Z)-Zotiraciclib (hydrochloride) | 1321626-25-8 | C23H25ClN4O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15166A
Encompass_Preferred
(E/Z)-Zotiraciclib ((E/Z)-TG02) is an orally active inhibitor of CDK2, JAK2, and FLT3 with IC50s of 13, 73, and 56 nM, respectively. It effectively inhibits the proliferation of cancer cells and can be used for cancer research.
  • Potent inhibitor of CDK2, JAK2, and FLT3
  • Effectively inhibits proliferation of cancer cells
  • Can be used for cancer research
  • Inhibits CDK2 biomarker pRb
  • Demonstrates tumor growth inhibition in animal models
Catalog No. 50-003-75986