Learn More
Cayman Chemical E Z2propyl2PentenoIc Ac 10mg
SDP
Supplier:  Cayman Chemical 1959110 MG
Encompass_Preferred
A mixture of (E)-2-propyl-2-pentenoic acid and (Z)-2-propyl-2-pentenoic acid and an active metabolite of valproic acid; (E)-2-propyl-2-pentenoic acid increases the seizure threshold in the MEST test in mice at 100 mg/kg
Catalog No. 50-298-4459