Learn More
TRANSNETYX INC E18 Mouse Cortex Complete Culturing Kit
SDP
Supplier:  TRANSNETYX INC KTC57ECX
Encompass
Complete Culturing Kit The Complete Culturing Kit contains everything you need to start a culture from fresh tissue. E18 tissue is shipped in 2ml of THRYVE Embryonic Storage Media Complete with B27. Use tissue immediately for highest cell yield however, tissue can be stored for one week at 4-8°C. This kit is perfect for the researcher just starting out or for those who want reliability. Great for classroom education allowing a hands-on approach for the next generation researchers.Details: E18 C57 Mouse Cortex Complete Culturing Kit Includes: 1 pair E18 Mouse Cortex 12ml NbActiv1 6.2mg Papain 5ml HE-Ca 1 Fire Polished Pipette with rubber bulb 1 pair PDL Coated Coverslips
Catalog No. NC3718316