Learn More
ABclonal Technology E2F8 Rabbit pAb
SDP
Supplier:  ABclonal Technology A113520UL
Encompass_Preferred
This gene encodes a member of a family of transcription factors which regulate the expression of genes required for progression through the cell cycle. The encoded protein regulates progression from G1 to S phase by ensuring the nucleus divides at the proper time. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Catalog No. 50-001-14315