Learn More
Medchemexpress LLC E3 ligase ligand 32 | 2300099-98-1 | MFCD34595287 | 99.9% | 338.16 g·mol⁻1 | C13H12BrN3O3 | 10 G

Supplier: Medchemexpress LLC HYW38203810G
E3 ligase Ligand 32 is a small-molecule ligand for E3 ubiquitin ligases supplied for research use. The compound is accompanied by analytical documentation and is available in multiple pack sizes, including a 10 g option. It is intended for biochemical and cellular studies of ubiquitination and targeted protein degradation; follow safety and handling guidance provided by the manufacturer.
- Small-molecule ligand for E3 ubiquitin ligase.
- Supplied with certificate of analysis and datasheet.
- High purity 99.9%.
- Molecular formula C13H12BrN3O3; molecular weight 338.16 g·mol⁻1.
- Available in multiple pack sizes, including 10 g.
- Intended for biochemical and cellular research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.