Learn More
Medchemexpress LLC E3 ligase Ligand 63 | 2304754-51-4 | 99.8% | C13H12BrN3O3 | 10 G
Supplier: Medchemexpress LLC HY4551210G
E3 ligase Ligand 63 is an E3 ligase ligand used for synthesizing CDK2 degrader 2. It functions as part of PROTACs, which are molecules designed to harness the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
- Utilized for synthesizing CDK2 degrader 2
- Part of PROTACs, which incorporate two distinct ligands
- Operates by harnessing the intracellular ubiquitin-proteasome system
- Achieves selective degradation of target proteins