Learn More
Medchemexpress LLC E3 ligase Ligand 64 | 2760850-50-6 | ≥98.0% | C₁₂H₁₀BrFN₄O₂ | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY49385
Encompass_Preferred
E3 ligase Ligand 64 is a ligand for E3 ligase, used for the synthesis of RP03707. This product is a solid with a molecular weight of 341.14 and a chemical formula of C₁₂H₁₀BrFN₄O₂. It is intended for laboratory research use only and should be handled by qualified scientists in appropriately equipped facilities. It is classified with several hazards, including acute oral toxicity, skin and eye irritation, and potential respiratory irritation.
  • Identified as 2,4(1H,3H)-Pyrimidinedione, 1-(6-bromo-5-fluoro-1-methyl-1H-indazol-3-yl)dihydro-.
  • Used for synthesis.
  • For research use only.
  • Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping at room temperature if less than 2 weeks.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Harmful if swallowed.
Catalog No. 50-003-58503