Learn More
Cayman Chemical E3 LIgase LIgnd 2 1g
SDP
Supplier:  Cayman Chemical 366641 G
Encompass_Preferred
An active metabolite of thalidomide and a PROTAC synthetic intermediate; decreases production of TNF-α induced by TPA in THP-1 cells at 30 µM; completely inhibits the proliferation HUVECs at 333 µM; inhibits VEGF- and FGF2-induced angiogenesis by 14% in a CAM assay in utero at 100 UG/embryo
Catalog No. 50-291-3849