Learn More
Cayman Chemical E3 LIgndLInkR Conjugate 3 25mg
SDP
Supplier:  Cayman Chemical 3666625 MG
Encompass_Preferred
A PROTAC building block containing thalidomide; has been used in the synthesis of PROTACs for the degradation ALK and BET family of proteins with anticancer activity
Catalog No. 50-291-0010